15 COMPOSES DANS ACETONITRILE (Ampoules de 1 ml ) Acetaldehyde-DNPH [CAS:1019-57-4] 101.5mg/l Acetone-DNPH [CAS:1567-89-1] 82mg/l Benzaldehyde-DNPH [CAS:1157-84-2] 53.9mg/l n-Butyraldehyde-DNPH [CAS:1527-98-6] 69.9mg/l Crotonaldehyde-DNPH [CA 1 * 5 Ampoul
Catalog # CMPLF98C.56.N.1
Supplier: CUSTOM MADE CHEMICALS LAB
Frequently Bought Together