Spanner wrenches
Tricoise, pour écrous Ø 65 - 75 mm, pour colonnes HiScale™ 26, Acier galvanisé
Catalog # 28-9647-77
Supplier: Cytiva
Jump to:
Product Details & Documents
These spanner wrenches are used with HiScale™ columns. To readjust the position of the adapter in the column a spanner can be used to turn the end cap. To prevent the column from rotating, a second spanner can be used to hold the end housing.
Documents
Cytiva CyDye Licensing Statements Cytiva General Licensing Statements
Additional Product Details
Specifications
- Désignation:Tricoise, pour écrous Ø 65 - 75 mm, pour colonnes HiScale™ 26
- Matière:Acier galvanisé
- Seq:02