Saisie de commande
Belgium
Lien pour commandesContactUsLinkComponent
Acide nitrique 32,5%

Acide nitrique 32,5%

Bidon Plastique, 10 L
Catalog # PRLSMC305295.410
Corrosive
Acide nitrique 32,5%
Catalog # PRLSMC305295.410
Supplier:  
CAS Number:  7697-37-2
Acide nitrique 32,5%

Bidon Plastique, 10 L

CAS Number:  7697-37-2
Discontinued
Product has been discontinued. However we found similar alternative products.
Alternatives
ACIDE NITRIQUE 32,5% (W/W) 1 * 5 L Catalog #PRLSMC305295.5000

Product PRLSMC305295.5000 is a replacement for PRLSMC305295.410 but varies by Brand, Packaging or Quantity.

Product Details & Documents
Synonyms:  Nitric acid solution
  • CAS Number:7697-37-2
  • Formula:HNO₃
  • MDL Number:MFCD00011349
  • Molecular Weight:63.01 g/mol
  • Boiling Pt:∼86 °C (1013 hPa)
  • Melting Pt:–42 °C
  • Density:1,2…1.4 g/cm³ (20 °C)
  • Storage Temperature:Température ambiante
  • EINECS:231-714-2
  • UN:2031
  • ADR:8,II
  •  :Density: 1.51 g/cm³ (20 °C)
Specifications

Specifications

  • Type de conditionnement:
    Bidon Plastique
  • Cdt:
    10 L

Specifications