Saisie de commande
Belgium
Lien pour commandesContactUsLinkComponent
Human Recombinant Rab3A (from E. coli)

Human Recombinant Rab3A (from E. coli)

N° de catalogue ABCAAB109964-50
Fournisseur:   Abcam
Human Recombinant Rab3A (from E. coli)
N° de catalogue ABCAAB109964-50
Fournisseur:  Abcam
Numéro CAS:  
Human Recombinant Rab3A (from E. coli)
Human Recombinant Rab3A (from E. coli)
Numéro CAS:  
Prix
544,00 €
Connexion Pour la disponibilité
1 * 50 µG
Product Details & Documents

Recombinant Human Rab3A protein is a Human Full Length protein, in the 1 to 220 aa range, expressed in Escherichia coli, with >90% purity and suitable for SDS-PAGE, MS.

Attention: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Spécifications

Spécifications

  • Formulaire:
    Liquid
  • Activité biologique:
    Standard
  • Purity:
    >90%
  • Synonyme de protéine:
    Ras-related protein Rab-3A
  • Conjugaison:
    Unconjugated
  • Concentration:
    0,5 mg/ml
  • Source:
    E. coli
  • Cat. No.:
    ABCAAB109964-50
  • Conditions de stockage:
    Store at −20 °C
  • Formulation:
    pH: 8; Constituents: 20% Glycerol (glycerin, glycerine), 0,316% Tris HCl, 0,058% Sodium chloride, 0,0154% (R*,R*)-1,4-Dimercaptobutan-2,3-diol
  • Exempt de substance animale:
    N
  • Tested applications:
    MS, SDS-PAGE
  • Species:
    Human
  • Nom de la protéine/du peptide:
    Rab3A
  • Type de protéine/peptide:
    Recombinant
  • Température de transport:
    20

Spécifications

Recommandations

Les recommandations seront personnalisées en fonction de vos préférences d’achat uniquement si vous avez donné votre consentement en activant l’option " Améliorer mon expérience d’achat " sur la page "Mon compte".

Dans le cas contraire, vous recevrez des recommandations génériques.