Pycnometers for bituminous materials
Pycnometer with ring mark, matt shield and numbering, borosilicate glass 3.3, 1000 ml, 60
N° de catalogue NEUB0321-51-1000
Fournisseur: NEUBERT VOLUME GLASSWAERE
Pycnometers for bituminous materials
N° de catalogue NEUB0321-51-1000
Fournisseur: NEUBERT VOLUME GLASSWAERE
Aller à :
Product Details & Documents
Soda-lime glass or borosilicate glass 3.3.
- Two-part assembly both parts with identification number
- Cap with funnel for ease of filling
- Ring mark on cap stem
Spécifications
- RN:60
- Volume nominal:1000 ml
- Désignation:Pycnometer with ring mark, matt shield and numbering, borosilicate glass 3.3
- Seq:03