Saisie de commande
Belgium
Lien pour commandesContactUsLinkComponent
Dissolved oxygen meter, handheld, Orion Star™ A123

Dissolved oxygen meter, handheld, Orion Star™ A123

Handheld DO meter Orion Star A123, instrument alone
N° de catalogue 664-0143
Fournisseur:   Thermo Orion
Dissolved oxygen meter, handheld, Orion Star™ A123
N° de catalogue 664-0143
Fournisseur:  Thermo Orion
Dissolved oxygen meter, handheld, Orion Star™ A123
Dissolved oxygen meter, handheld, Orion Star™ A123

Handheld DO meter Orion Star A123, instrument alone

Prix
830,00 €
Connexion Pour la disponibilité
1 * 1 ST
Product Details & Documents

The Orion Star™ A123 dissolved oxygen handheld meter offers easy operation for basic analysis in the field. Ideal for taking routine dissolved oxygen and temperature measurements.

  • Measure dissolved oxygen in percent saturation or mg/L with temperature using polarographic DO probes
  • Calibrate using water-saturated air or custom value (Winkler titration)
  • Automatically compensate DO readings using ATC temperature, manual salinity input and manual barometric pressure
  • Simplified, easy-to-read display shows main measurement with temperature, stability indicator and electrode condition icon
  • User-friendly operation with basic calibration text prompts
  • Easily recall calibration data for procedure checks
  • Non-volatile memory preserves 50 point internal data log and meter settings, even with loss of power
  • IP 67-rated housing is waterproof for rugged field operation

Accepts Thermo Scientific™ Orion™ polarographic dissolved oxygen probes.

The included electrode stand can be used to easily place sensors in samples and simplify storage.

Certifications: CE, TUV 3-in-1 and FCC Class A

Livraison: Supplied with literature CD, printed quick start guide, meter test certificate and 3-year replacement warranty. The kit additionally includes the 083005MD Orion polarographic DO probe (1,5 m cable) with calibration sleeve, 080513 DO probe maintenance kit, protective meter armor with electrode holder and hard-sided field case.

Spécifications
  • Désignation:
    Handheld DO meter Orion Star A123, instrument alone
  • Seq:
    01
  • Alimentation:
    AC adapter or 4 AA batteries
  • Gamme de température:
    –5,0...+50,0 °C
  • Acquisition des données - Capacité:
    50
  • Langues:
    English
  • Oxygène dissous - Type de sonde:
    Polarographic
  • Modèle:
    A123
  • Exactitude de la températue:
    ±0,1 °C
  • Connexions:
    9-pin mini-DIN
  • Affichage:
    LCD
  • Température de service:
    5...45 °C (ambient)
  • Acquisition des données - Enregistrement:
    Manual / Automatic with AUTO-READ™
  • Détermination dela température:
    0,1 °C
  • Conformités:
    IP 67
  • Batterie - Durée de vie:
    2000 h
  • Poids:
    0,5 kg
  • Oxygène dissous - saturation:
    0 - 200 %
  • L×P×H:
    100×240×650 mm
  • OD Concentration:
    0 - 20 mg/L
  • Plage d'humidité de fonctionnement:
    5 - 85%, non-condensing
  • Cat. No.:
    664-0143
  • Oxygène dissous - Facteur de salinité:
    Manual, 0 - 45 ppt
  • Mémoire:
    Non-volatile memory preserves log and settings
  • Oxygène dissous - Gamme de pression barométrique:
    Manual, 450 - 850 mmHg
  • OD Etalonnage:
    Water-saturated air, manual (Winkler)
  • OD Précision:
    ±0,2%
  • Oxygène dissous - Résolution:
    0,1 / 0,01 mg/L
  • Nombre de canaux:
    1
Accessories

Informations sur le produit

  • N° de catalogue
    Désignation
    Disponibilité
    Prix
  • 662-1547
    Adaptateur universel supplémentaire de rechange pour appareils de mesure de paillasse et portables Thermo Scientific™ Orion™ Star A et appareils de mesure hérités Orion™ Star et Star Plus
    Connexion Pour la disponibilité
    1 * 1 ST - 89,80 €

Informations sur le produit

  • N° de catalogue
    Désignation
    Pour
    Disponibilité
    Prix
  • ORIOSTARA-CS
    Sacoche de terrain rigide pour appareils de mesure portables, contenant un appareil de mesure (avec ou sans blindage), des supports d'électrodes portables, des piles supplémentaires, des solutions, une électrode de pH dans une gaine de stockage, une sonde de conductivité et une sonde à oxygène dissous dans une gaine de stockage
    Orion Star A120, A220 + A320
    Connexion Pour la disponibilité
    1 * 1 ST - 179,00 €

Informations sur le produit

  • N° de catalogue
    Désignation
    Pour
    Disponibilité
    Prix
  • ORIO1210005
    Sacoche de terrain souple, comprenant des séparateurs intérieurs et des poches extérieures qui permettent d'organiser l'appareil de mesure et les accessoires sur le terrain
    Orion Star A120, A220 + A320, Orion Star et Star Plus
    Connexion Pour la disponibilité
    1 * 1 ST - 200,00 €

Informations sur le produit

  • N° de catalogue
    Désignation
    Pour
    Disponibilité
    Prix
  • ORIOSTARA-AR
    Blindage de protection pour appareils de mesure portables, avec dragonne et support intégré, gaine de stockage d'électrode de pH, support d'électrode de pH, et support de capteur de conductivité et d'oxygène dissous
    Orion Star A320, A220 + A120
    Connexion Pour la disponibilité
    1 * 1 ST - 127,00 €

Informations sur le produit

  • N° de catalogue
    Désignation
    Pour
    Disponibilité
    Prix
  • ORIOSTARA-ESCD
    Conductimètre portable Orion™ Star A et support de sonde d'oxygène dissous, pouvant être fixé de n'importe quel côté du blindage de l'appareil de mesure portable, et pouvant accueillir des sondes de conductivité de 12/15 mm de Ø et une sonde à oxygène dissous lorsqu'elle est placée dans sa gaine d'étalonnage
    Orion Star A320, A220 + A120
    Connexion Pour la disponibilité
    1 * 1 ST - 36,60 €

Recommandations

Les recommandations seront personnalisées en fonction de vos préférences d’achat uniquement si vous avez donné votre consentement en activant l’option " Améliorer mon expérience d’achat " sur la page "Mon compte".

Dans le cas contraire, vous recevrez des recommandations génériques.