Tubing sinkers, fits tubing with inner Ø 5,5 - 6,2 mm
These Teflon® sinkers help to keep tubing at the bottom of the container.
#: 83639.320
: VWR Chemicals
#: 20864.320
#: 612-1681
: VWR Collection
#: 20063.365
#: 112-2371
#: 612-3700
#: 112-2756
#: 83640.320
#: 115-0600
#: 112-4847
: Ansell
#: 1.00030.2500
: Merck
#: 1.09535.0001
#: 291-0057
: HEATHROW
#: 23595.328
#: 31770.294
#: 32096.291
#: 83645.320
#: 1.09439.1000
#: 112-4195
#: 613-3508
: EPPENDORF