Erlenmeyer flasks with screw caps
2000 ml, 280 mm, 166 mm, GL 45
N° de catalogue WITG5.507.007
Fournisseur: witeg Labortechnik
Aller à :
Product Details & Documents
Erlenmeyer flasks, made of borosilicate glass, with screw-thread, graduated can be used for the storage of culture media and mixing.
- Excellent chemical resistance
- High temperature resistance
- Minimal thermal expansion, giving relatively high resistance to temperature changes
Spécifications
- Ø ext.:166 mm
- Filetage:GL 45
- Capacité:2000 ml
- Hauteur:280 mm