Adapters, receiver, plain bent, Quickfit®
19/26 NS, 24/29 NS
N° de catalogue BIBBRA9/23
Fournisseur:   SciLabware
Adapters, receiver, plain bent, Quickfit®
N° de catalogue BIBBRA9/23
Fournisseur:  SciLabware
Adapters, receiver, plain bent, Quickfit®
Adapters, receiver, plain bent, Quickfit®
19/26 NS, 24/29 NS
Prix
206,00 €
Connexion Pour la disponibilité
1 * 5 ST
Product Details & Documents
Receiver, delivery, PYREX® borosilicate glass
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
Plain bend without vent.
Spécifications
  • Rodage mâle:
    24/29 NS
  • Rodage femelle:
    19/26 NS
  • Seq:
    03