Pipette tips, Ovation®
VistaStak™, 1250 µl, -, 3 rack layers, 192 each
N° de catalogue VIST4060-9026
Fournisseur:   VISTALAB TECHNOLOGIES
Pipette tips, Ovation®
N° de catalogue VIST4060-9026
Fournisseur:  VISTALAB TECHNOLOGIES
Pipette tips, Ovation®
Pipette tips, Ovation®
VistaStak™, 1250 µl, -, 3 rack layers, 192 each
Prix
79,00 €
Connexion Pour la disponibilité
1 * 576 ST
Product Details & Documents
Ideal for use with Ovation and MLA pipettes.
  • Made of 100% pure virgin PP
  • Provide a perfect fit
  • Deliver reliable performance
  • Autoclavable
Spécifications
  • Emballage:
    3 rack layers, 192 each
  • Stérile:
    -
  • Volume:
    1250 µl
  • Seq:
    09
  • Style de pointe:
    VistaStak™