Extraction thimbles
Extraction thimbles
25×60 mm, 1,0 mm
N° de catalogue WHAT10350416
Extraction thimbles
N° de catalogue WHAT10350416
Fournisseur:  
Extraction thimbles
Extraction thimbles
25×60 mm, 1,0 mm
Arrêté
Product Details & Documents
Grade 603 standard cellulose extraction thimbles, made of high quality cellulose, are available in a range of sizes suitable for use in thimble holders and automated extraction apparatus to ensure optimal fit.
Spécifications
  • Ø int.×h:
    25×60 mm
  • Seq:
    01
  • Épaisseur:
    1,0 mm