Cleaning solution 6 g Na4-EDTA + 65 g NaOH/l for determination of o-PO4 and total P (FIA/CFA) EN ISO 15681-1:2004, DEV D45 EN ISO 15 681-2:2004, DEV D46... 1 * 1 L
N° de catalogue KRAF19412.3000
Fournisseur: Bernd Kraft
Frequently Bought Together