Safety caps
Safety cap II with 1 basic cap, PP (blue) with PTFE cone, 2 ports (includes 2 PFA fittings, 3,2 mm (blue and red), 1 venting valve, including air filter, 1 μm, PTFE membrane), for NS29/32 ground neck bottles
N° de catalogue HEWL5043-0221
Fournisseur: Agilent
Aller à :
Product Details & Documents
These safety caps have been designed for optimal sealing with an integrated exhaust valve providing pressurisation during solvent extraction and allowing proper solvent flow to your HPLC system.
- Caps rotate freely, preventing tube twisting during bottle exchange
- Constructed of PTFE and PFA for high chemical resistance
The exhaust valve contains a PTFE membrane to prevent contamination of your solvents from dirt and dust particles.
Spécifications
- Désignation:Safety cap II with 1 basic cap, PP (blue) with PTFE cone, 2 ports (includes 2 PFA fittings, 3,2 mm (blue and red), 1 venting valve, including air filter, 1 μm, PTFE membrane), for NS29/32 ground neck bottles
- Seq:02
Accessories
Informations sur le produit
- N° de catalogueDésignationDisponibilitéPrix
Informations sur le produit
- N° de catalogueDésignationDisponibilitéPrix
Informations sur le produit
- N° de catalogueDésignationDisponibilitéPrix
Informations sur le produit
- N° de catalogueDésignationDisponibilitéPrix