Safety caps
Safety caps
Safety cap II with 1 basic cap, PP (blue) with PTFE cone, 2 ports (includes 2 PFA fittings, 3,2 mm (blue and red), 1 venting valve, including air filter, 1 μm, PTFE membrane), for NS29/32 ground neck bottles
N° de catalogue HEWL5043-0221
Fournisseur:   Agilent
Safety caps
N° de catalogue HEWL5043-0221
Fournisseur:  Agilent
Safety caps
Safety caps
Safety cap II with 1 basic cap, PP (blue) with PTFE cone, 2 ports (includes 2 PFA fittings, 3,2 mm (blue and red), 1 venting valve, including air filter, 1 μm, PTFE membrane), for NS29/32 ground neck bottles
Prix
Demander un prix
Connexion Pour la disponibilité
1 * 1 ST
Product Details & Documents
These safety caps have been designed for optimal sealing with an integrated exhaust valve providing pressurisation during solvent extraction and allowing proper solvent flow to your HPLC system.
  • Caps rotate freely, preventing tube twisting during bottle exchange
  • Constructed of PTFE and PFA for high chemical resistance
The exhaust valve contains a PTFE membrane to prevent contamination of your solvents from dirt and dust particles.
Spécifications
  • Désignation:
    Safety cap II with 1 basic cap, PP (blue) with PTFE cone, 2 ports (includes 2 PFA fittings, 3,2 mm (blue and red), 1 venting valve, including air filter, 1 μm, PTFE membrane), for NS29/32 ground neck bottles
  • Seq:
    02
Accessories
Informations sur le produit
  • N° de catalogue
    Désignation
    Disponibilité
    Prix
  • HEWL5043-0229
    Fittings, PFA, for 1,6 mm tube
    Connexion Pour la disponibilité
    1 * 5 ST
Informations sur le produit
  • N° de catalogue
    Désignation
    Disponibilité
    Prix
  • HEWL5043-0238
    Collector, 3 ports
    Connexion Pour la disponibilité
    1 * 1 ST
Informations sur le produit
  • N° de catalogue
    Désignation
    Disponibilité
    Prix
  • HEWL5043-0239
    Adapter, for 2×3,2 mm tubes
    Connexion Pour la disponibilité
    1 * 1 ST
Informations sur le produit
  • N° de catalogue
    Désignation
    Disponibilité
    Prix
  • HEWL5043-0255
    Fittings, PFA, for 3,2 mm tube
    Connexion Pour la disponibilité
    1 * 6 ST