Soxhlet extractors
Soxhlet extractors
Cône 29/32, 500 ml, 60/46 NS
N° de catalogue WITG2.212.500
Fournisseur:   witeg Labortechnik
Soxhlet extractors
N° de catalogue WITG2.212.500
Fournisseur:  witeg Labortechnik
Soxhlet extractors
Soxhlet extractors
Cône 29/32, 500 ml, 60/46 NS
Prix
152,00 €
Connexion Pour la disponibilité
1 * 1 ST
Product Details & Documents
Borosilicate glass. For extraction from solids with solvents.
  • Very good chemical resistance
  • Very good temperature resistance
The solvent is recirculated continuously and distilled from the extracted sample before contacting the solid again. The extract is concentrated in the distillation flask. The method allows an effective separation with relatively small amounts of extracting solvents.
Spécifications
  • Capacité extracteur:
    500 ml
  • Rodage femelle:
    60/46 NS
  • Désignation:
    Cône 29/32
  • Seq:
    01