Saisie de commande
Belgium
Lien pour commandesContactUsLinkComponent
Microtubes à centrifuger

Microtubes à centrifuger

0,4 ml, naturel, With attached sealing caps, Non graduated, conical bottom, non sterile
N° de catalogue RATI5615400
Fournisseur:   RATIOLAB
Microtubes à centrifuger
N° de catalogue RATI5615400
Fournisseur:  RATIOLAB
Microtubes à centrifuger
Microtubes à centrifuger

0,4 ml, naturel, With attached sealing caps, Non graduated, conical bottom, non sterile

Prix
Demander un prix
Connexion Pour la disponibilité
1 * 1.000 ST
Product Details & Documents

PP, translucent.

  • Autoclavable with opened cap at 121 °C, 20 min
  • Natural colour or different colours for comfortable identification
  • Tight sealing cap
  • Safe and easy to be opened and closed

With or without flat cap.

Spécifications
  • Type:
    With attached sealing caps
  • Version:
    Non graduated, conical bottom, non sterile
  • Capacité:
    0,4 ml
  • Couleur:
    naturel
  • Seq:
    01
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.