Bandage mull
Gauze, 20-ply, zigzag layers, 80 cm wide, 40 m long
N° de catalogue HART2064072
Fournisseur: HARTMANN PAUL
Aller à :
Product Details & Documents
Mull for making individual bandages and wound dressings.
- 20 threads
- Laid 8-fold in zigzag layers to a width of 10 cm
- Good purity and absorbency
Certifications: EN 14079
Spécifications
- Désignation:Gauze, 20-ply, zigzag layers, 80 cm wide, 40 m long