Accessories for storage tubes, barcoded
96× storage tube screw cap, with O-Ring, on mat, clear with cover, orange, 96 caps per mat
N° de catalogue 731-0242
Fournisseur: Corning
Aller à :
Product Details & Documents
Documents
Certificats
Rechercher par numéro de lotSpécifications
- Emballage:96 caps per mat
- Couleur:orange
- Désignation:96× storage tube screw cap, with O-Ring, on mat, clear with cover