Disposable cannulas
Disposable cannulas, G22, noir, 50 mm, +, 0,7×50 mm
Catalog # EHRS15884650
Supplier: EHRHARDT SOEHNE
Jump to:
Product Details & Documents
Disposable cannulas.
- Pyrogen-free, not toxic
- With lancet cut
- EO-sterilisation
Documents
Certificates
Search by Lot NumberSpecifications
- Ø×L:0,7×50 mm
- Jauge:G22
- Stérile:+
- Couleur:noir
- Désignation:Disposable cannulas
- Longueur:50 mm
- Seq:04