D(+)-Glucose monohydraté
Glass bottle for solids, 250 g
  XXXX76020761.0250
 :   Chemicals
D(+)-Glucose monohydraté
  XXXX76020761.0250
 :  Chemicals
 :  14431-43-7
D(+)-Glucose monohydraté
D(+)-Glucose monohydraté
Glass bottle for solids, 250 g
 :  14431-43-7
1 * 250 g
This product has been blocked by your organization. Please contact your purchasing department for more information
 :  (2S,3R,4S,5S,6R)-6-(Hydroxymethyl)tetrahydro-2H-pyran-2,3,4,5-tetraol hydrate, Dextrose monohydrate
  •  :14431-43-7
  •  :C₆H₁₂O₆·H₂O
  •  :MFCD00149450
  •  :198.17 g/mol
  •  :83 °C
  •  :Température ambiante
  •  :200-075-1
  • Type de conditionnement:
    Glass bottle for solids
  • Cdt:
    250 g