D(+)-Glucose monohydraté
Glass bottle for solids, 250 g
XXXX76020761.0250
: (2S,3R,4S,5S,6R)-6-(Hydroxymethyl)tetrahydro-2H-pyran-2,3,4,5-tetraol hydrate, Dextrose monohydrate
- :14431-43-7
- :C₆H₁₂O₆·H₂O
- :MFCD00149450
- :198.17 g/mol
- :83 °C
- :Température ambiante
- :200-075-1
- Type de conditionnement:Glass bottle for solids
- Cdt:250 g