Sleeves for ground joints, rigid
14/20
N° de catalogue COWI020.114.1
Fournisseur: COWIE
Aller à :
Product Details & Documents
Made of PTFE, grease-free, high vacuum full length rigid sleeves for standard glass joints, with substantial shoulder for ease of handling and repeated use.
- Rigid sleeves and adapters are inert
- Usable to 280˚C high vacuum
- The leakage rate is less
- High vacuum performance is obtained by transverse sealing rings
Documents
Spécifications
- Pour rodage:14/20
- Seq:02