Sleeves for ground joints, rigid
Sleeves for ground joints, rigid
19/22
N° de catalogue COWI020.119.1
Fournisseur:   COWIE
Sleeves for ground joints, rigid
N° de catalogue COWI020.119.1
Fournisseur:  COWIE
Sleeves for ground joints, rigid
Sleeves for ground joints, rigid
19/22
Prix
87,40 €
Connexion Pour la disponibilité
1 * 10 ST
Product Details & Documents
Made of PTFE, grease-free, high vacuum full length rigid sleeves for standard glass joints, with substantial shoulder for ease of handling and repeated use.
  • Rigid sleeves and adapters are inert
  • Usable to 280˚C high vacuum
  • The leakage rate is less
  • High vacuum performance is obtained by transverse sealing rings
Spécifications
  • Pour rodage:
    19/22
  • Seq:
    03