Sterilisation adhesive tapes
Indicator adhesive tapes, for non-woven paper, 24 mm
SPSA31BL2550VEU
Marketsource
: SPS
These adhesive tapes are adapted for all types of sterilisation.
- The dispenser can be used for easier cutting tape
- These products do not contain any latex or heavy metals
- Largeur:24 mm
- Désignation:Indicator adhesive tapes, for non-woven paper
- Seq:04