Sterilisation adhesive tapes
Indicator adhesive tapes, for non-woven paper, 24 mm
  SPSA31002550VE
Marketsource
 :   SPS
Sterilisation adhesive tapes
  SPSA31002550VE
 :  SPS
Marketsource
Sterilisation adhesive tapes
Sterilisation adhesive tapes
Indicator adhesive tapes, for non-woven paper, 24 mm
6,79 €
1 * 50 m
These adhesive tapes are adapted for all types of sterilisation.
  • The dispenser can be used for easier cutting tape
  • These products do not contain any latex or heavy metals
  • Largeur:
    24 mm
  • Désignation:
    Indicator adhesive tapes, for non-woven paper
  • Seq:
    03