Sterilisation adhesive tapes
Indicator adhesive tapes, for non-woven paper, 18 mm
Catalog # SPSA31001950VE
Marketsource
Supplier: SPS
Jump to:
Product Details & Documents
These adhesive tapes are adapted for all types of sterilisation.
- The dispenser can be used for easier cutting tape
- These products do not contain any latex or heavy metals
Documents
SPSA31001950VE_TDS_EN
Datasheet
Specifications
- Largeur:18 mm
- Désignation:Indicator adhesive tapes, for non-woven paper
- Seq:01