Sterilisation adhesive tapes
Sterilisation adhesive tapes
Indicator adhesive tapes, for non-woven paper, 18 mm
Catalog # SPSA31001950VE
Marketsource
Supplier:   SPS
Sterilisation adhesive tapes
Catalog # SPSA31001950VE
Supplier:  SPS
Marketsource
Sterilisation adhesive tapes
Sterilisation adhesive tapes
Indicator adhesive tapes, for non-woven paper, 18 mm
Price
5,41 €
Log in for availability
1 * 1 ROU
Product Details & Documents
These adhesive tapes are adapted for all types of sterilisation.
  • The dispenser can be used for easier cutting tape
  • These products do not contain any latex or heavy metals
Documents
Specifications
  • Largeur:
    18 mm
  • Désignation:
    Indicator adhesive tapes, for non-woven paper
  • Seq:
    01