Waste caps
Waste caps
Waste cap, includes O-ring, 1/4" nuts, ferrules and plug, B 83, 4 ports with 1/4-28 thread for 1/8" or 1/16" tubing, 1 port with 1/2-20 thread for 1/4" tubing, PP
N° de catalogue JOURJR-S-18304
Fournisseur:   VICI JOUR RESEARCH
Waste caps
N° de catalogue JOURJR-S-18304
Fournisseur:  VICI JOUR RESEARCH
Waste caps
Waste cap, includes O-ring, 1/4" nuts, ferrules and plug, B 83, 4 ports with 1/4-28 thread for 1/8" or 1/16" tubing, 1 port with 1/2-20 thread for 1/4" tubing, PP
Prix
575,00 €
Connexion Pour la disponibilité
1 * 1 ST
Product Details & Documents
These waste caps include three or four ports for ¹/₈ and ¹/₁₆" tubing. Two versions of each waste cap are available, with an extra port to accommodate either a 1/2-20 port for ¹/₄" O.Ø tubing, or a barbed hose adapter.
Ordering information: Barbed adapters must be ordered separately.
Spécifications
  • Filetage:
    B 83
  • Désignation:
    Waste cap, includes O-ring, 1/4" nuts, ferrules and plug
  • Matière:
    PP
  • Seq:
    03
  • Raccord:
    4 ports with 1/4-28 thread for 1/8" or 1/16" tubing, 1 port with 1/2-20 thread for 1/4" tubing