Waste caps
Waste cap, includes O-ring, 1/4" nuts, ferrules and plug, S 61, 3 ports with 1/4-28 thread for 1/8" or 1/16" tubing, 1 port with 1/2-20 thread for 1/4" tubing, PP
N° de catalogue JOURJR-S-16103
Fournisseur: VICI JOUR RESEARCH
Aller à :
Product Details & Documents
These waste caps include three or four ports for ¹/₈ and ¹/₁₆" tubing. Two versions of each waste cap are available, with an extra port to accommodate either a 1/2-20 port for ¹/₄" O.Ø tubing, or a barbed hose adapter.
Ordering information: Barbed adapters must be ordered separately.
Spécifications
- Filetage:S 61
- Désignation:Waste cap, includes O-ring, 1/4" nuts, ferrules and plug
- Matière:PP
- Seq:12
- Raccord:3 ports with 1/4-28 thread for 1/8" or 1/16" tubing, 1 port with 1/2-20 thread for 1/4" tubing