Extender tools
Extender tool, with 1/4"hex drive, Aluminium, Standard knurl
Catalog # UPCHP-291
Supplier: UPCHURCH SCIENTIFIC
Jump to:
Product Details & Documents
These tools made from aluminium or Delrin® can be used to tighten most knurled nuts in hard to reach places. For precise tightening, the extender tools listed with ¼" hex drives are designed to adapt to any torque wrench with a female ¼" socket.
Specifications
- Désignation:Extender tool, with 1/4"hex drive
- Matière:Aluminium
- Seq:01
- À utiliser avec une tête de type:Standard knurl