Hardened fine scissors
Sharp/sharp, curved tips, 90 mm, 24 mm
FSCI14091-09
Marketsource
These scissors are made of stainless steel.
- Type:Sharp/sharp, curved tips
- Longueur:90 mm
- Seq:04
- Longueur de lame:24 mm
These scissors are made of stainless steel.