Fine scissors, ToughCut®
Sharp/sharp, curved tips, Toughcut®, 115 mm, 25 mm, +
FSCI14059-11
Marketsource
These scissors are made of stainless steel.
- Type:Sharp/sharp, curved tips, Toughcut®
- Longueur:115 mm
- Seq:03
- Serrated:+
- Longueur de lame:25 mm



