DNA ladder, O'RangeRuler™
O'RangeRuler 5 bp DNA Ladder, range 10 to 100 bp, ready to use, 50 µg, 1 KIT
N° de catalogue SM1303
Fournisseur:   Thermo Fisher Scientific
DNA ladder, O'RangeRuler™
N° de catalogue SM1303
Numéro CAS:  
DNA ladder, O'RangeRuler™
DNA ladder, O'RangeRuler™
O'RangeRuler 5 bp DNA Ladder, range 10 to 100 bp, ready to use, 50 µg, 1 KIT
Numéro CAS:  
Prix
191,00 €
Connexion Pour la disponibilité
1 * 1 KIT
Product Details & Documents
O'RangeRuler™ DNA ladders are step ladders that consist of basic unit repeats of 10, 15, 20, 50, 100, 200, or 500 bp. The ladders have a higher density of bands at a specific DNA size range and are therefore suitable for more accurate sizing of DNA in agarose or non denaturing polyacrylamide gels.
  • Step ladders - ligation products of DNA fragments of equal increments
  • Sharp bands, evenly spaced reference bands
  • Ready to use - premixed with loading dye for direct loading
  • Room temperature storage
  • Supplied with loading dye for sample DNA
 : Supplied with 6X orange DNA loading dye for sample DNA.
Spécifications
Spécifications
  • Cdt:
    1 KIT
  • Désignation:
    O'RangeRuler 5 bp DNA Ladder, range 10 to 100 bp, ready to use, 50 µg