Accessories for round tissue float baths, TFB
Accessory set includes thermometer (0 - 100 °C), thermometer support and aluminium lid, Model TFB 55
N° de catalogue MEDT11-5500-00
Fournisseur: MEDITE
Aller à :
Product Details & Documents
Spécifications
- Désignation:Accessory set includes thermometer (0 - 100 °C), thermometer support and aluminium lid
- Seq:08
- Pour:Model TFB 55