Screw caps for micro tubes
Screw caps for micro tubes, naturel
N° de catalogue SARS65.716
Marketsource
Fournisseur: SARSTEDT
Aller à :
Product Details & Documents
PP, coloured.
- Excellent chemical resistance
- Excellent temperature resistance
Screw cap with O-ring seal.
Spécifications
- Couleur:naturel
- Désignation:Screw caps for micro tubes
- Seq:06