Accessories for glove boxes, Precise™
Accessories for glove boxes, Precise™
Digital hygrometer, handheld, measures both percent relative humidity from 0-100% and temperature in ° F/° C; incl. 9 volt alkaline battery
N° de catalogue LBCP5244800
Fournisseur:   LABCONCO
Accessories for glove boxes, Precise™
N° de catalogue LBCP5244800
Fournisseur:  LABCONCO
Accessories for glove boxes, Precise™
Accessories for glove boxes, Precise™
Digital hygrometer, handheld, measures both percent relative humidity from 0-100% and temperature in ° F/° C; incl. 9 volt alkaline battery
Prix
2 440,00 €
Connexion Pour la disponibilité
1 * 1 ST
Product Details & Documents
Spécifications
  • Désignation:
    Digital hygrometer, handheld, measures both percent relative humidity from 0-100% and temperature in ° F/° C; incl. 9 volt alkaline battery
  • Seq:
    15