Dissolution vessels, QLA
Dissolution vessels, QLA
QLA amber glass dissolution vessel, TruCenter, serialised, 1000 ml
N° de catalogue QLAAGLA9AM-VK
Fournisseur:   QUALITY LAB ACCESSORIES
Dissolution vessels, QLA
N° de catalogue QLAAGLA9AM-VK
Dissolution vessels, QLA
Dissolution vessels, QLA
QLA amber glass dissolution vessel, TruCenter, serialised, 1000 ml
Prix
Demander un prix
Connexion Pour la disponibilité
1 * 1 ST
Product Details & Documents
Dissolution vessels are extremely important because the vessels associated with various dissolution baths are not dimensionally the same. Replacing a vessel with one not specifically designed for the instrument could impair vessel centering.
  • Made of high quality borosilicate glass or plastic
  • Dissolution vessels comply with both USP (711) and dimensional specifications
Also larger or smaller vessel inside diameters can change hydrodynamics. Both variables could impact dissolution rates.
Spécifications
  • Désignation:
    QLA amber glass dissolution vessel, TruCenter, serialised, 1000 ml