Bucket, Rectangular
Seau de 25 l, gris, 22 L
Catalog # 129-0454
Supplier: FHP VILEDA SCS
Jump to:
Product Details & Documents
Polypropylene bucket with stainless steel handle. For use on the CE Bucket Trolley or the CE pre-prepared trolley.
- Liquid measurement lines molded inside of bucket
- Color coding clips included to allow differentiation in colours of buckets (red, blue, green, yellow)
- Autoclavable
Highest performance for the best contamination control.
Specifications
- Capacité:22 L
- Couleur:gris
- Désignation:Seau de 25 l
- Seq:02