Cell viability assay kits
Cell viability assay kits
MTT Cell Viability Assay kit, 1000 assays, 1 KIT
N° de catalogue 30006.
Fournisseur:   Biotium
Cell viability assay kits
N° de catalogue 30006.
Fournisseur:  Biotium
Numéro CAS:  
Cell viability assay kits
MTT Cell Viability Assay kit, 1000 assays, 1 KIT
Numéro CAS:  
Prix
256,00 €
Connexion Pour la disponibilité
1 * 1 KIT
Product Details & Documents
MTT, XTT, and resazurin are reduced by mitochondrial metabolic activity to yield coloured or fluorescent products, and thus are useful for and assaying cell viability and quantitating cell number. MTT and XTT are reduced to coloured formazin salts that can be measured by absorbance. MTT generates an insoluble formazin salt, requiring cell lysis before the absorbance can be measured, while XTT does not require cell lysis for measurement. Resazurin is a non-fluorescent blue dye that is reduced to the pink fluorescent compound resorufin, which can be measured by fluorescence or absorbance.
Documents
Spécifications
Spécifications
  • Cdt:
    1 KIT
  • Désignation:
    MTT Cell Viability Assay kit, 1000 assays
Spécifications
Recommandations
Les recommandations seront personnalisées en fonction de vos préférences d’achat uniquement si vous avez donné votre consentement en activant l’option " Améliorer mon expérience d’achat " sur la page "Mon compte".
Dans le cas contraire, vous recevrez des recommandations génériques.