VWR® Basic, Frocks, SMS
bleu, XL
Supplier: VWR Collection
Jump to:
Product Details & Documents
These SMS frocks are manufactured from a latex-free, multi-layer spunbonded - melt-blown - spunbonded (SMS) fabric, that offers high tensile strength while remaining soft, comfortable and breathable.
- Snap collar, raglan sleeves, snap closure
- Fluid and particulate barrier
- Light chemical splash resistance
Colour: White or blue
Specifications
- Taille de confection:XL
- Couleur:bleu
- Seq:09
- Cat. No.:113-1175
- Type de fermeture:Avec boutons pression
- Applications des tenues:Salle propre|Laboratoire
- Genre:Unisex
- Poches:sans poches