Erlenmeyer flasks, narrow neck, with baffles
Erlenmeyer flasks, narrow neck, with baffles
100 ml
N° de catalogue NEUB0749-10-0100
Erlenmeyer flasks, narrow neck, with baffles
N° de catalogue NEUB0749-10-0100
Erlenmeyer flasks, narrow neck, with baffles
100 ml
Prix
84,50 €
Connexion Pour la disponibilité
1 * 1 ST
Product Details & Documents
Borosilicate glass 3.3.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
Ideal for use in the preparation of cultures.
Spécifications
  • Capacité:
    100 ml
  • Seq:
    01