Microsampling tools
Micro specimen tweezers
Catalog # PERCN9302609
Supplier: PerkinElmer
Jump to:
Product Details & Documents
This microsampling tool kit is suitable for FTIR microscopy.
Delivery information: Microsampling tool kit includes dissecting forceps, roller knife, straight micro probe, aluminum probe handle, pin vise.
Specifications
- Désignation:Micro specimen tweezers
- Seq:03
- Cat. No.:PERCN9302609