Closures for headspace vials
Closures for headspace vials
Blue PTFE coated silicone steel magnetic cap, pre-assembled
N° de catalogue PERCN6356559
Fournisseur:   PerkinElmer
Closures for headspace vials
N° de catalogue PERCN6356559
Fournisseur:  PerkinElmer
Closures for headspace vials
Blue PTFE coated silicone steel magnetic cap, pre-assembled
Prix
139,00 €
Connexion Pour la disponibilité
1 * 1 ST
Product Details & Documents
Caps for 20 ml glass headspace crimp top round bottom vials which have a 23 mm OD and 75,5 mm height.
Spécifications
  • Désignation:
    Blue PTFE coated silicone steel magnetic cap, pre-assembled
  • Seq:
    11