Reservoir system
Reservoir, full, 150 ml, +
Supplier: Beckman Coulter
Jump to:
Product Details & Documents
This modular reservoir system holds liquids for pipetting with a manual pipettor or liquid handling systems including the full suite of Biomek liquid handlers.
- Half and quarter reservoirs must be used with a frame
- Full reservoirs hold up to 150 ml of liquid, and do not require a frame
- Each frame can accommodate up to four quarter reservoirs or two half reservoirs
- All reservoirs are sterile and made of PP
Not for use with 96 and 384 multi channel pipetting heads.
Documents
Certificates
Search by Lot NumberSpecifications
- Stérile:+
- Capacité:150 ml
- Désignation:Reservoir, full
- Seq:03