Reservoir system
Reservoir system
Reservoir, full, 150 ml, +
Catalog # BECL372784
Marketsource
Supplier:   Beckman Coulter
Reservoir system
Catalog # BECL372784
Supplier:  Beckman Coulter
Marketsource
Reservoir system
Reservoir system
Reservoir, full, 150 ml, +
Price
Inquire For Price
Log in for availability
1 * 12 ST
Product Details & Documents
This modular reservoir system holds liquids for pipetting with a manual pipettor or liquid handling systems including the full suite of Biomek liquid handlers.
  • Half and quarter reservoirs must be used with a frame
  • Full reservoirs hold up to 150 ml of liquid, and do not require a frame
  • Each frame can accommodate up to four quarter reservoirs or two half reservoirs
  • All reservoirs are sterile and made of PP
Not for use with 96 and 384 multi channel pipetting heads.
Documents
Specifications
  • Stérile:
    +
  • Capacité:
    150 ml
  • Désignation:
    Reservoir, full
  • Seq:
    03