Round canisters
4 L, naturel, B83, Standard, 155×155×338 mm
Catalog # SCTV107712
Supplier: S.C.A.T.
Jump to:
Product Details & Documents
SCAT round canisters are ideal for collecting and disposing of smaller quantities of solvents due to their compact dimensions. Several SCAT round canisters can be easily stationed at the HPLC system and equipped with SCAT SafeyWasteCaps.
- Made of either robust HDPE or PP and therefore resistant to all solvents commonly used in the laboratory
- Available in the common threads S65, S70/71 and B83
- Filling capacities range from very compact 2 to 4 L, 10, 25, 50 and 60 L for the collection of voluminous solvents
- Many individual features such as carrying handles, handles or measuring scales, which ensure safe handling of the hazardous substances
SCAT round canisters are a perfect alternative or supplement to conventional SCAT canisters to provide physical separation of solvents and prevent mixing of hazardous liquids.
Documents
GHS Poster Important Lab and HPLC Safety Guidelines
Application Notes
Specifications
- Filetage:B83
- Type:Standard
- Capacité:4 L
- Couleur:naturel
- Seq:02
- l×p×h:155×155×338 mm
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



