Sodium hydroxyde solution 50% (p/p), BAKER ANALYZED®, J.T.Baker®
Jerrican, poly pail, 19 L
Fournisseur: Avantor
Sodium hydroxyde solution 50% (p/p), BAKER ANALYZED®, J.T.Baker®
N° de catalogue 3727-07
Fournisseur: Avantor
Numéro CAS: 1310-73-2
Aller à :
Product Details & Documents
MPORTANT: Material will freeze if stored below 60°F (16°C).
: IMPORTANT: Material will freeze if stored below 60°F (16°C). Protect from air to avoid absorption of carbon dioxide.
Synonymes: Caustic soda
- Numéro CAS:1310-73-2
- Formule:NaOH
- Numéro MDL:MFCD00003548
- Poids moléculaire:40 g/mol
- Point d'ébullition:119…135 °C (1013 hPa)
- Point de fusion:9…10 °C
- Densité:1,53 g/cm³ (20 °C)
- Température de stockage:Température ambiante
- EINECS:215-185-5
- UN:1824
- ADR:8,II
Spécifications
Résultats des tests de spécification
- For Laboratory,Research,or Manufacturing Use:50 - 52%
- Protect from air to avoid absorption of carbon dioxide.:≤20
- Assay (NaOH) (by acidimetry):50 - 52 %
- Color (APHA):≤20
- Sodium Carbonate (Na₂CO₃):≤0.1 %
- Chloride (Cl):≤0.002 %
- Sulfate (SO₄):≤0.001 %
- Ammonium Hydroxide Precipitate:≤0.01 %
- Silica (SiO₂), For Information Only:≤5 ppm
- Heavy Metals (as Ag):≤0.001 %
- Potassium (K):≤0.01 %
- Nitrogen Compounds (as N):≤5 ppm
- Phosphate (PO₄):≤5 ppm
- Trace Impurities - Iron (Fe):≤5 ppm
- Trace Impurities - Nickel (Ni):≤5.000 ppm
Spécifications
- Type de conditionnement:Jerrican, poly pail
- Cdt:19 L
Spécifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



