Staining dishes
Staining dishes
Staining set for haematology
Catalog # BOPT10-20
Supplier:   BIO OPTICA
Staining dishes
Catalog # BOPT10-20
Supplier:  BIO OPTICA
Staining dishes
Staining dishes
Staining set for haematology
Price
240,00 €
Log in for availability
1 * 3 ST
Product Details & Documents
These staining dishes are made of PET (polyethylene terephthalate) stiffened with glass fibres. They are resistant against solvents especially in cytology and histology staining processes.
  • Resist temperature between 0 to 170 °C
  • Can be used with immunohistochemistry procedures
Specifications
  • Désignation:
    Staining set for haematology
  • Seq:
    03
Accessories
Product Information
  • Catalog #
    Désignation
    Availability
    Price
  • BOPT10-42
    Rack for 25 slides with plastic handle
    Log in for availability
    1 * 6 ST - 96,00 €
Recommandations
Les recommandations seront personnalisées en fonction de vos préférences d’achat uniquement si vous avez donné votre consentement en activant l’option " Améliorer mon expérience d’achat " sur la page "Mon compte".
Dans le cas contraire, vous recevrez des recommandations génériques.