Staining dishes
Staining set for haematology
Catalog # BOPT10-20
Supplier: BIO OPTICA
Jump to:
Product Details & Documents
These staining dishes are made of PET (polyethylene terephthalate) stiffened with glass fibres. They are resistant against solvents especially in cytology and histology staining processes.
- Resist temperature between 0 to 170 °C
- Can be used with immunohistochemistry procedures
Specifications
- Désignation:Staining set for haematology
- Seq:03
Accessories
Product Information
- Catalog #DésignationAvailabilityPrice