Staining dishes
Staining dishes
Staining dish with attached lid
N° de catalogue BOPT10-30
Fournisseur:   BIO OPTICA
Staining dishes
N° de catalogue BOPT10-30
Fournisseur:  BIO OPTICA
Staining dishes
Staining dishes
Staining dish with attached lid
Prix
243,00 €
Connexion Pour la disponibilité
1 * 12 ST
Product Details & Documents
These staining dishes are made of PET (polyethylene terephthalate) stiffened with glass fibres. They are resistant against solvents especially in cytology and histology staining processes.
  • Resist temperature between 0 to 170 °C
  • Can be used with immunohistochemistry procedures
Spécifications
  • Désignation:
    Staining dish with attached lid
  • Seq:
    01
Accessories
Informations sur le produit
  • N° de catalogue
    Désignation
    Disponibilité
    Prix
  • BOPT10-42
    Rack for 25 slides with plastic handle
    Connexion Pour la disponibilité
    1 * 6 ST - 96,00 €