Staining dishes
Staining dish with attached lid
N° de catalogue BOPT10-30
Fournisseur: BIO OPTICA
Aller à :
Product Details & Documents
These staining dishes are made of PET (polyethylene terephthalate) stiffened with glass fibres. They are resistant against solvents especially in cytology and histology staining processes.
- Resist temperature between 0 to 170 °C
- Can be used with immunohistochemistry procedures
Spécifications
- Désignation:Staining dish with attached lid
- Seq:01
Accessories
Informations sur le produit
- N° de catalogueDésignationDisponibilitéPrix