Blood collection needles, BD Vacutainer® Eclipse™
21, 32 mm, vert
N° de catalogue 613-3110
Aller à :
Arrêté
Le produit a été arrêté. Cependant, nous avons trouvé des produits alternatifs similaires.
Alternatives
AIGUILLE VACUTAINER ECLIPSE 21G 1,25INCH 1 * 48 Pce N° de catalogueBDAM368609
Product BDAM368609 is een vervanging voor 613-3110, maar verschilt per merk, verpakking of hoeveelheid.
Product Details & Documents
The BD Vacutainer® Eclipse™ blood collection needle is a safety-engineered, multi-sample blood collection needle that offers a simple, effective way to collect blood while reducing the possibility of needlestick injuries. It features a safety shield that allows for one-handed activation to cover the needle immediately upon withdrawal from the vein and confirms proper activation with an audible click.
- Built-in safety shield is aligned with the bevel of the needle
- The safety shield permanently locks into place, significantly reducing the risk of needlestick injuries
- The pre-attached holder (only supplied with some models) minimises the risk of accidental needlestick injuries from the holder end of the device
Spécifications
- Code couleur:vert
- Jauge:21
- Longueur:32 mm
- Seq:02
Aanbevelingen
Aanbevelingen worden alleen gepersonaliseerd op basis van uw winkelvoorkeuren als u daarvoor toestemming hebt gegeven door Verbeter mijn winkelervaring" op Mijn Account pagina" aan te zetten.
Anders ontvangt u algemene aanbevelingen.