Powder funnels
120 mm
N° de catalogue HECH44002044
Fournisseur: GLASWARENFABRIK KARL HECHT
Aller à :
Product Details & Documents
Powder funnels, made of PP, translucent. For decanting powdery materials. Also suitable for liquids. Cone angle 60°.
- Excellent chemical resistance
- Excellent temperature resistance
- Autoclavable
Documents
HECH44002044_TDS_EN
Fiche technique
Spécifications
- Ø ext. entonnoir sup.:120 mm