Racks for freezer
17 (1×17), 32 mm, 140×140×625 mm
Catalog # RATI5400038
Supplier: RATIOLAB
Jump to:
Product Details & Documents
For the storing of cryoboxes in freezer cabinets and chests.
- Robust construction
- Handles for convenient transport
- Made of stainless steel with surfaces for perfect cleaning
Delivery information: Cryo racks are supplied with shelf sizes for any standard cryoboxes.
Specifications
- Compartiments:17 (1×17)
- l×p×h ext.:140×140×625 mm
- Seq:16
- Hauteur de cryoboîte:32 mm



