Saisie de commande
Belgium
Lien pour commandesContactUsLinkComponent
RNAspin Mini Kits

RNAspin Mini Kits

Kit illustra™ RNAspin Mini Isolation, 50 prép., 50 Tests
N° de catalogue 25-0500-71
Fournisseur:   Cytiva
RNAspin Mini Kits
N° de catalogue 25-0500-71
Fournisseur:  Cytiva
Numéro CAS:  
RNAspin Mini Kits
RNAspin Mini Kits

Kit illustra™ RNAspin Mini Isolation, 50 prép., 50 Tests

Numéro CAS:  
Prix
323,00 €
Connexion Pour la disponibilité
1 * 50 Tests
Product Details & Documents

RNAspin Mini RNA Isolation Kit is a complete RNA purification kit designed for rapid extraction of high-quality total RNA from a wide range of sample types.

  • High-quality output RNA from diverse sample types
  • Recovers high-quality total RNA since genomic DNA is removed via on-column DNase I treatment
  • For maximum yield and purity, prefilters are included to reduce lysate viscosity
  • Results can be obtained with even small amounts of sample
  • Lysis buffer is less susceptible to foaming to ensure valuable RNA sample is not wasted
  • Simple and convenient format suitable for all levels of expertise

RNA extraction is the purification of RNA from biological samples. It is complicated by the ubiquitous presence of ribonuclease enzymes in cells and tissues, which can rapidly degrade RNA, since it is less stable than DNA.

Besides other methods in the RNA isolation protocol to help counter RNA degradation, the equipment used for RNA preparation is usually kept separate from other lab equipment and treated (or has been pre-treated) with chemicals that can destroy RNases, such as diethyl pyrocarbonate (DEPC). RNAspin Mini Isolation Kit can be used to isolate total RNA from cultured cells, tissue, bacteria, or yeast. The isolated RNA is of a high enough quality and quantity (up to 100 μg) for multiple downstream experiments and sensitive enzymatic applications like RT-PCR, real time PCR, primer extension, RNAseq, and RNase protection assays.
The RNAspin Mini Kit has been used for the isolation of high-quality total RNA from several types of source material including hard-to-lyse bacterial strains and can be applied in downstream experiments such as quantitative reverse transcriptase PCR (QRT-PCR), Northern blot, and microarray experiments. The kit is supplied with a full RNA extraction protocol booklet with a detachable, quick reference protocol card, and all the necessary components including prefilters and DNase I.

Livraison: The kit is supplied with a full protocol booklet with a detachable, quick reference protocol card, and all the necessary components including prefilters and DNase I.

Spécifications

Spécifications

  • Cdt:
    50 Tests
  • Désignation:
    Kit illustra™ RNAspin Mini Isolation, 50 prép.
  • Cat. No.:
    25-0500-71
  • Prep size:
    Mini
  • Refine target molecule:
    Total RNA
  • Target molecule:
    RNA
  • Method / Format:
    Column (silica-based)
  • Sample type:
    Cultured cells|Human/animal cells and tissues
  • Désignation:
    illustra™ RNAspin Mini Isolation Kit

Spécifications

Recommandations

Les recommandations seront personnalisées en fonction de vos préférences d’achat uniquement si vous avez donné votre consentement en activant l’option " Améliorer mon expérience d’achat " sur la page "Mon compte".

Dans le cas contraire, vous recevrez des recommandations génériques.