Saisie de commande
Belgium
Lien pour commandesContactUsLinkComponent
PIG® absorbent pillows

PIG® absorbent pillows

Super pillows, PP skin/cellulose filler, 53 L, 14 pillows/box, 200×510×40 mm
N° de catalogue NEWPPIL205
Fournisseur:   NEW PIG
PIG® absorbent pillows
N° de catalogue NEWPPIL205
Fournisseur:  NEW PIG
PIG® absorbent pillows
PIG® absorbent pillows

Super pillows, PP skin/cellulose filler, 53 L, 14 pillows/box, 200×510×40 mm

Prix
113,00 €
Connexion Pour la disponibilité
1 * 14 ST
Product Details & Documents

Add extra absorbency to your spill cleanup efforts with high-capacity PIG Pillows. Also great for steady leaks and drips.

  • Pillow has large surface area, high capacity and fast-wicking filler to quickly soak up liquids
  • Polypropylene skin resists tearing; reduces dust and holds in liquid, even when saturated
  • Cellulose filler is highly absorbent for long-term use or big spill cleanup
  • Compact design is great for catching persistent drips and leaks in small spaces or adding absorbency for spill response
  • Absorbs and retains oils, coolants, solvents and water; not recommended for acids, caustics or other corrosive liquids

85% mixture of pre- and post-consumer recycled cellulose filler. Can be incinerated after use to reduce waste or for fuels blending.

Spécifications
  • Désignation:
    Super pillows, PP skin/cellulose filler
  • Seq:
    03
  • l×p×h:
    200×510×40 mm
  • Capacité d'absorption:
    53 L
  • Emballage:
    14 pillows/box
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.